ENA448969551 Enamine

1-benzyl-1,3-diazinan-2-one

  • MDL number MFCD11870028
  • Cas Number : 34790-80-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 161-165
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-215180

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-215180
Availability details for
Quantity
  • Availability
  • Add to Cart