ENA448969552 Enamine

methyl 2-(hydroxymethyl)-4-methyl-1,3-thiazole-5-carboxylate

  • MDL number MFCD26798636
  • Cas Number : 1565712-91-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-215182

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-215182
Availability details for
Quantity
  • Availability
  • Add to Cart