ENA448969553 Enamine

5-{2-azabicyclo[2.2.1]heptan-2-yl}-2-methylaniline

  • MDL number MFCD17248094
  • Cas Number : 1340189-20-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-215185

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-215185
Availability details for
Quantity
  • Availability
  • Add to Cart