ENA448969558 Enamine

5-fluoro-3-(piperidin-4-yl)-1H-indole

  • MDL number MFCD08669442
  • Cas Number : 149669-43-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-213536

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-213536
Availability details for
Quantity
  • Availability
  • Add to Cart