ENA448969560 Enamine

2-(butylamino)-5-nitrobenzene-1-sulfonamide

  • MDL number MFCD28505959
  • Cas Number : 1286648-18-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-213538

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-213538
Availability details for
Quantity
  • Availability
  • Add to Cart