ENA448969562 Enamine

1-chlorobutane-2,3-dione

  • MDL number MFCD18806712
  • Cas Number : 5559-62-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-215210

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-215210
Availability details for
Quantity
  • Availability
  • Add to Cart