ENA448969565 Enamine

2-[2-(methoxymethyl)-1,3-thiazol-4-yl]acetonitrile

  • MDL number MFCD17251043
  • Cas Number : 1343362-21-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-215213

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-215213
Availability details for
Quantity
  • Availability
  • Add to Cart