ENA448969569 Enamine

bis(N-butylguanidine); sulfuric acid

  • MDL number MFCD00035582
  • Cas Number : 3867-74-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 206-207
Purity 95
Salt Data 2H2SO4
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-215224

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-215224
Availability details for
Quantity
  • Availability
  • Add to Cart