ENA448969570 Enamine

N-methyl-4-(piperidin-3-yloxy)pyridine-2-carboxamide dihydrochloride

  • MDL number MFCD28505960
  • Cas Number : 1803562-51-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-213542

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-213542
Availability details for
Quantity
  • Availability
  • Add to Cart