ENA448969572 Enamine

2-[1-(1-methyl-1H-imidazol-2-yl)ethoxy]acetic acid

  • MDL number MFCD20658582
  • Cas Number : 1378848-47-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-213544

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-213544
Availability details for
Quantity
  • Availability
  • Add to Cart