ENA448969579 Enamine

2-[(4-methyl-1,3-thiazol-5-yl)methoxy]acetic acid

  • MDL number MFCD18432905
  • Cas Number : 1265892-62-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-213545

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-213545
Availability details for
Quantity
  • Availability
  • Add to Cart