ENA448969580 Enamine

1-(bromomethyl)-4-cyclopentylbenzene

  • MDL number MFCD11110251
  • Cas Number : 1260851-03-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-215287

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-215287
Availability details for
Quantity
  • Availability
  • Add to Cart