ENA448969582 Enamine

1,3-dimethyl-1H-indazol-6-amine

  • MDL number MFCD18432764
  • Cas Number : 221681-92-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-215294

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-215294
Availability details for
Quantity
  • Availability
  • Add to Cart