ENA448969589 Enamine

1-[3-(aminomethyl)pyrrolidin-1-yl]-2,2-dimethylpropan-1-one

  • MDL number MFCD16667651
  • Cas Number : 1339164-60-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-215333

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-215333
Availability details for
Quantity
  • Availability
  • Add to Cart