ENA448969592 Enamine

N-methyl-2-(propan-2-yl)aniline

  • MDL number MFCD09999703
  • Cas Number : 7117-18-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-215339

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-215339
Availability details for
Quantity
  • Availability
  • Add to Cart