ENA448969594 Enamine

1-(2-methoxypyridin-4-yl)ethan-1-amine

  • MDL number MFCD13188773
  • Cas Number : 1060807-28-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-213550

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-213550
Availability details for
Quantity
  • Availability
  • Add to Cart