ENA448969666 Enamine

2-iodo-N,N-dimethylacetamide

  • MDL number MFCD01670617
  • Cas Number : 73664-43-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-216063

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-216063
Availability details for
Quantity
  • Availability
  • Add to Cart