ENA448969755 Enamine

4-chloro-1,3-dimethyl-1H-pyrazole

  • MDL number MFCD09743376
  • Cas Number : 15878-44-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-216170

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-216170
Availability details for
Quantity
  • Availability
  • Add to Cart