ENA448969820 Enamine

2-chloro-N-(1-cyanocyclobutyl)acetamide

  • MDL number MFCD09042619
  • Cas Number : 926273-74-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-47217

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-47217
Availability details for
Quantity
  • Availability
  • Add to Cart