ENA448969854 Enamine

tert-butyl N-[2-(piperidin-2-yl)ethyl]carbamate

  • MDL number MFCD04038512
  • Cas Number : 885954-19-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-76325

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-76325
Availability details for
Quantity
  • Availability
  • Add to Cart