ENA448969864 Enamine

[(2S)-2-aminopropyl]dimethylamine

  • MDL number MFCD19204342
  • Cas Number : 70831-55-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84875

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-84875
Availability details for
Quantity
  • Availability
  • Add to Cart