ENA448969875 Enamine

4,4-dimethylcyclohexane-1-carbonitrile

  • MDL number MFCD20319755
  • Cas Number : 25186-25-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-90286

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-90286
Availability details for
Quantity
  • Availability
  • Add to Cart