ENA448969877 Enamine

tert-butyl 4-cyanopiperazine-1-carboxylate

  • MDL number MFCD09029184
  • Cas Number : 113534-02-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 101-102
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-91723

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-91723
Availability details for
Quantity
  • Availability
  • Add to Cart