ENA448969883 Enamine

5-(trifluoromethyl)pyrimidin-2-amine

  • MDL number MFCD10696951
  • Cas Number : 69034-08-8
  • Molecular Weight : 163.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-(trifluoromethyl)-2-pyrimidinamine
Inchi Code 1S/C5H4F3N3/c6-5(7,8)3-1-10-4(9)11-2-3/h1-2H,(H2,9,10,11)
Inchi Key OUVVCECMELPNNT-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-94954

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-94954
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart