ENA455585226 Enamine

(1S)-3-methyl-1-{[1,2,4]triazolo[4,3-a]pyridin-3-yl}butan-1-amine dihydrochloride

  • MDL number MFCD28714519
  • Cas Number : 1807939-67-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-206537
Availability details for
Quantity
  • Availability
  • Add to Cart