ENA455585264 Enamine

2-[3-(pyrimidin-5-yl)propyl]-2,3-dihydro-1H-isoindole-1,3-dione

  • MDL number MFCD28714569
  • Cas Number : 1803607-85-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-214219

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-214219
Availability details for
Quantity
  • Availability
  • Add to Cart