ENA455585310 Enamine

3-amino-8-methoxy-3,4-dihydro-2H-1-benzopyran-4-one hydrochloride

  • MDL number MFCD28714674
  • Cas Number : 22406-62-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-216270

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-216270
Availability details for
Quantity
  • Availability
  • Add to Cart