ENA455585352 Enamine

2-fluorocyclooctane-1-sulfonyl chloride

  • MDL number MFCD28714747
  • Cas Number : 1803589-48-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-217094

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-217094
Availability details for
Quantity
  • Availability
  • Add to Cart