ENA455585356 Enamine

(2S)-2-[(5-bromopyridin-2-yl)amino]-3-methylbutan-1-ol hydrochloride

  • MDL number MFCD28714758
  • Cas Number : 1807940-77-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-217275
Availability details for
Quantity
  • Availability
  • Add to Cart