ENA455585364 Enamine

tert-butyl N-(4-oxopentyl)carbamate

  • MDL number MFCD27957633
  • Cas Number : 197358-56-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-206879
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart