ENA455585369 Enamine

methyl 5-amino-4-methylthiophene-2-carboxylate

  • MDL number MFCD27952446
  • Cas Number : 1023655-30-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Product Link https://www.enaminestore.com/catalog/EN300-207129

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207129
Availability details for
Quantity
  • Availability
  • Add to Cart