ENA455585385 Enamine

methyl 5-bromo-4-oxopentanoate

  • MDL number MFCD18800850
  • Cas Number : 53856-93-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. 0

Safety Information

msds https://www.enaminestore.com/catalog/EN300-114360
Availability details for
Quantity
  • Availability
  • Add to Cart