ENA455585429 Enamine

4-{6-[(3-hydroxypropyl)amino]imidazo[1,2-b]pyridazin-3-yl}phenol

  • MDL number MFCD26392215
  • Cas Number : 1012344-09-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-210734
Availability details for
Quantity
  • Availability
  • Add to Cart