ENA455585444 Enamine

4-[(cyclopropylamino)methyl]-3-fluorobenzonitrile hydrochloride

  • MDL number MFCD28714550
  • Cas Number : 1803607-36-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-211706

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-211706
Availability details for
Quantity
  • Availability
  • Add to Cart