ENA455585481 Enamine

methyl (3S)-3-amino-3-(naphthalen-1-yl)propanoate hydrochloride

  • MDL number MFCD24422437
  • Cas Number : 1245606-67-0
  • Molecular Weight : 265.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-213222

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-213222
Availability details for
Quantity
  • Availability
  • Add to Cart