ENA455585490 Enamine

ethyl 2-(6-chloropyridazin-3-yl)acetate

  • MDL number MFCD09881204
  • Cas Number : 1023817-10-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-213419

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-213419
Availability details for
Quantity
  • Availability
  • Add to Cart