ENA455585499 Enamine

methyl 3-[(1,3-dioxaindan-5-yl)amino]butanoate

  • MDL number MFCD12429800
  • Cas Number : 1155159-50-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-213660

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-213660
Availability details for
Quantity
  • Availability
  • Add to Cart