ENA455585511 Enamine

2-bromo-5-(bromomethyl)furan

  • MDL number MFCD16860578
  • Cas Number : 251649-67-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. -20

Safety Information

msds https://www.enaminestore.com/catalog/EN300-139605
Availability details for
Quantity
  • Availability
  • Add to Cart