ENA455585563 Enamine

2-(4-bromo-2-chlorophenyl)-2-hydroxy-3-(1H-1,2,4-triazol-1-yl)propanoic acid hydrochloride

  • MDL number MFCD28714577
  • Cas Number : 85792-59-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-214718

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-214718
Availability details for
Quantity
  • Availability
  • Add to Cart