ENA455585571 Enamine

2,6-dimethylpiperidin-4-one hydrochloride

  • MDL number MFCD11656348
  • Cas Number : 1005397-62-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-214805

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-214805
Availability details for
Quantity
  • Availability
  • Add to Cart