ENA455585577 Enamine

4-fluoro-2-methylbenzene-1-carbothioamide

  • MDL number MFCD09757564
  • Cas Number : 1256482-73-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-214923
Availability details for
Quantity
  • Availability
  • Add to Cart