ENA455585602 Enamine

(1R,10S)-tricyclo[8.3.1.0,3,8]tetradeca-3,5,7-trien-14-one

  • MDL number MFCD28714590
  • Cas Number : 1807940-74-4
  • Molecular Weight : 200.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-215166

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-215166
Availability details for
Quantity
  • Availability
  • Add to Cart