ENA455585629 Enamine

2-(5-chlorothiophen-2-yl)-6-methoxyquinoline-4-carboxylic acid

  • MDL number MFCD05176330
  • Cas Number : 897565-19-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-215331

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-215331
Availability details for
Quantity
  • Availability
  • Add to Cart