ENA455585658 Enamine

3-bromo-1-(4-ethylphenyl)propan-1-one

  • MDL number MFCD18391261
  • Cas Number : 33994-10-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. 0

Safety Information

msds https://www.enaminestore.com/catalog/EN300-215445
Availability details for
Quantity
  • Availability
  • Add to Cart