ENA455585677 Enamine

6-bromo-4-tert-butyl-7-fluoro-1,2,3,4-tetrahydroquinoxalin-2-one

  • MDL number MFCD27429270
  • Cas Number : 1714857-13-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-215473

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-215473
Availability details for
Quantity
  • Availability
  • Add to Cart