ENA455585702 Enamine

6,8-dichloro-2-(5-chlorothiophen-2-yl)quinoline-4-carboxylic acid

  • MDL number MFCD05175596
  • Cas Number : 897558-65-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-215577

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-215577
Availability details for
Quantity
  • Availability
  • Add to Cart