ENA455585719 Enamine

[1-(3-aminopropyl)-1H-1,2,3-triazol-4-yl]methanol

  • MDL number MFCD16747742
  • Cas Number : 1132017-25-4
  • Molecular Weight : 156.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 72-75
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-215607

Safety Information

signalword Danger
pcodes P260,P264,P280,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P363,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-215607
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart