ENA455585746 Enamine

ethyl 3-(pyridin-2-yl)-1H-pyrazole-4-carboxylate

  • MDL number MFCD11865104
  • Cas Number : 1188263-92-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-215702

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-215702
Availability details for
Quantity
  • Availability
  • Add to Cart