ENA455585758 Enamine

[2-(dimethylamino)ethyl]({[4-(1H-imidazol-1-yl)phenyl]methyl})amine

  • MDL number MFCD12494244
  • Cas Number : 1152657-53-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-215714

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-215714
Availability details for
Quantity
  • Availability
  • Add to Cart