ENA455585786 Enamine

4-phenylcyclohexane-1-sulfonamide

  • MDL number MFCD16677696
  • Cas Number : 1249783-96-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-215779

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-215779
Availability details for
Quantity
  • Availability
  • Add to Cart